Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALGAPA1
Entrez Gene ID
253959
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
Chromosome Location
Chr.14: 35538352 - 35809304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283043.1 NP_001269972.1 23 - 24 3735 92
NM_001283044.1 NP_001269973.1 23 - 24 3837 92
NM_014990.1 NP_055805.1 22 - 23 3696 92
NM_194301.2 NP_919277.2 22 - 23 3696 92
XM_005267491.3 XP_005267548.1 23 - 24 3915 92
XM_005267492.3 XP_005267549.1 23 - 24 3915 92
XM_005267493.3 XP_005267550.1 22 - 23 3774 92
XM_005267498.4 XP_005267555.1 8 - 9 1525 92
XM_006720098.2 XP_006720161.1 23 - 24 5292 92
XM_006720099.2 XP_006720162.1 23 - 24 5292 92
XM_006720100.3 XP_006720163.1 22 - 23 5151 92
XM_006720101.2 XP_006720164.1 23 - 24 5292 92
XM_006720102.3 XP_006720165.1 23 - 24 4998 92
XM_006720103.2 XP_006720166.1 23 - 24 3915 92
XM_011536615.2 XP_011534917.1 23 - 24 5292 92
XM_011536616.2 XP_011534918.1 23 - 24 5292 92
XM_017021141.1 XP_016876630.1 22 - 23 5151 92
XM_017021142.1 XP_016876631.1 22 - 23 5151 92
XM_017021143.1 XP_016876632.1 23 - 24 5292 92
XM_017021144.1 XP_016876633.1 23 - 24 3915 92
XM_017021145.1 XP_016876634.1 22 - 23 3774 92
XM_017021146.1 XP_016876635.1 8 - 9 2961 92
XM_017021147.1 XP_016876636.1 8 - 9 1847 92
GenBank mRNA AB511280.1 - 23 - 24 3446 92
AY596970.1 - 22 - 23 3696 92
AY596971.1 - 22 - 23 3696 92
BC150596.1 - 23 - 24 3443 92

View Full Product Details