Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSHB
Entrez Gene ID
2488
Gene Name
follicle stimulating hormone beta subunit
Gene Aliases
HH24
Chromosome Location
Chr.11: 30231016 - 30235277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018080.1 NP_001018090.1 1 - 2 29 78

View Full Product Details