The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052967.1 | NP_443199.1 | 1 - 1 | 1050 | 77 |
GenBank mRNA | BC069127.1 | - | 1 - 1 | 1050 | 77 |
BC069345.1 | - | 1 - 1 | 1050 | 77 | |
BC095511.1 | - | 1 - 1 | 1115 | 77 | |
BC101176.2 | - | 1 - 1 | 1050 | 77 | |
BC101177.1 | - | 1 - 1 | 1050 | 77 |