The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_022007.2 | 1 - 2 | 68 | 97 | |
NR_046297.1 | 1 - 2 | 68 | 97 | ||
NR_046299.1 | 1 - 2 | 68 | 97 | ||
GenBank mRNA | DA885243.1 | - | 1 - 2 | 68 | 97 |