Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP41
Entrez Gene ID
286128
Gene Name
ZFP41 zinc finger protein
Gene Aliases
ZNF753, zfp-41
Chromosome Location
Chr.8: 143246821 - 143262705 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271156.2 NP_001258085.2 2 - 3 2582 53
GenBank mRNA BC034608.1 - 2 - 3 2582 53

View Full Product Details