The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007248.3 | NP_009179.2 | 2 - 2 | 1443 | 87 |
NM_016381.5 | NP_057465.1 | 1 - 1 | 1941 | 87 | |
NM_033629.4 | NP_338599.1 | 2 - 2 | 1621 | 87 | |
GenBank mRNA | AF151105.3 | - | 1 - 1 | 1598 | 87 |
AK315196.1 | - | 1 - 1 | 1231 | 87 | |
AL137745.1 | - | 1 - 1 | 1941 | 87 | |
BC019863.1 | - | 4 - 4 | 920 | 87 | |
BC023630.1 | - | 1 - 1 | 1187 | 87 | |
BT020052.1 | - | 1 - 1 | 775 | 87 | |
BT020053.1 | - | 1 - 1 | 775 | 87 |