Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX1-1
Entrez Gene ID
54729
Gene Name
NK1 homeobox 1
Gene Aliases
HSPX153, NKX-1.1, SAX2
Chromosome Location
Chr.4: 1402932 - 1406442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290079.1 NP_001277008.1 1 - 2 468 102

View Full Product Details