Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
79639
Gene Name
transmembrane protein 53
Gene Aliases
NET4
Chromosome Location
Chr.1: 44653247 - 44674608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300748.1 NP_001287677.1 2 - 3 400 73
GenBank mRNA AA490195.1 - 2 - 3 195 73
AK295356.1 - 1 - 2 93 73

View Full Product Details