The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110006.1 | 2 - 3 | 519 | 99 | |
GenBank mRNA | AK124669.1 | - | 2 - 3 | 527 | 99 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | fibroblast growth factor receptor 2 | FGFR2 | BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25 | -315.1kb | Sense | Chr. - 121478330 - 121598656 on Build GRCh38 |
Downstream | ATE1 antisense RNA 1 | ATE1-AS1 | 4.0kb | Anti-sense | Chr. - 121928312 - 121951965 on Build GRCh38 |