Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPS17
Entrez Gene ID
6218
Gene Name
ribosomal protein S17
Gene Aliases
DBA4, RPS17L, RPS17L1, RPS17L2, S17
Chromosome Location
Chr.15: 82536750 - 82540544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_111944.1 5 - 6 567 75
GenBank mRNA BM457623.1 - 4 - 5 460 75

View Full Product Details