The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_125372.1 | 1 - 2 | 121 | 145 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | isovaleryl-CoA dehydrogenase | IVD | ACAD2 | -5.2kb | Sense | Chr. - 40405485 - 40435948 on Build GRCh38 |
Downstream | carbohydrate sulfotransferase 14 | CHST14 | ATCS, D4ST1, EDSMC1, HNK1ST | 6.9kb | Sense | Chr. - 40470961 - 40473158 on Build GRCh38 |