Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CATIP-AS2
- Entrez Gene ID
-
103689911
- Gene Name
- CATIP antisense RNA 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 218326241 - 218357966 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125777.1 |
|
1 - 2 |
56 |
81 |
GenBank mRNA |
CD672221.1 |
- |
1 - 2 |
56 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ciliogenesis associated TTC17 interacting protein |
CATIP |
C2orf62 |
|
Anti-sense |
Chr. - 218356757 - 218368393 on Build GRCh38 |
Downstream |
CATIP antisense RNA 1 |
CATIP-AS1 |
|
8.7kb
|
Sense |
Chr. - 218366665 - 218367902 on Build GRCh38 |
View Full Product Details