The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301838.1 | NP_001288767.1 | 1 - 2 | 361 | 68 |
NR_126035.1 | 1 - 2 | 361 | 68 | ||
GenBank mRNA | AV751928.1 | - | - | 103 | 68 |
BE888740.1 | - | 1 - 2 | 52 | 68 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | RNA, U7 small nuclear 1 | RNU7-1 | RNU7, U7.1 | Sense | Chr. - 6943816 - 6943878 on Build GRCh38 | |
Downstream | protein tyrosine phosphatase, non-receptor type 6 | PTPN6 | HCP, HCPH, HPTP1C, PTP-1C, SH-PTP1, SHP-1, SHP-1L, SHP1 | 2.0kb | Sense | Chr. - 6946576 - 6961316 on Build GRCh38 |