Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCND1
Entrez Gene ID
595
Gene Name
cyclin D1
Gene Aliases
BCL1, D11S287E, PRAD1, U21B31
Chromosome Location
Chr.11: 69641105 - 69654474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK299044.1 - 3 - 4 474 79
AK313136.1 - 4 - 5 876 79
BC023620.2 - 4 - 5 854 79
BT019844.1 - 4 - 5 718 79
BT019845.1 - 4 - 5 718 79
M64349.1 - 4 - 5 862 79
X59798.1 - 4 - 5 865 79

View Full Product Details