Gene Transcripts

Gene Symbol
Phldb1
Entrez Gene ID
102693
Gene Name
pleckstrin homology-like domain, family B, member 1
Gene Aliases
AU041016, D330037A14Rik, Ll5a
Chromosome Location
Chr. 9: 44686308 - 44736151 on Build GRCm38
UniGene ID
Mm.28639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153537.4 NP_705765.3 6 660
GenBank mRNA AK122336.1 - 3 378
AK208267.1 - 2 223
BC058712.1 - 6 656 6842596

View Full Product Details