Gene Transcripts
- Gene Symbol
- Rtn1
- Entrez Gene ID
- 104001
- Gene Name
- reticulon 1
- Gene Aliases
- Nsp
- Chromosome Location
- Chr. 12: 72211749 - 72409054 on Build GRCm38
- UniGene ID
- Mm.221275
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007596.2 |
NP_001007597.1
|
5, 6 |
667 |
|
NM_001286448.1 |
NP_001273377.1
|
5, 6 |
1212 |
|
NM_153457.7 |
NP_703187.2
|
7, 8 |
2748 |
|
GenBank mRNA |
AB074899.1 |
- |
7, 8 |
2245 |
|
AK002997.1 |
- |
5, 6 |
665 |
|
AK013731.1 |
- |
5, 6 |
602 |
|
AK014358.1 |
- |
5, 6 |
620 |
|
AK132364.1 |
- |
7, 8 |
2748 |
|
BC027091.1 |
- |
5, 6 |
547 |
|
BC030455.1 |
- |
7, 8 |
2300 |
5401661 |
BC053926.1 |
- |
7, 8 |
2376 |
6312477 |
BC058579.1 |
- |
7, 8 |
2410 |
6490092 |
CT010392.1 |
- |
7, 8 |
2245 |
|
View Full Product Details