Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011084.2 | NP_035214.2 | 9 | 1272 | |
NM_207683.2 | NP_997566.1 | 4018 | |||
GenBank mRNA | AB008791.1 | - | 4018 | ||
AB008792.1 | - | 9 | 1267 | ||
AK054271.1 | - | 9 | 1148 | ||
BC150813.1 | - | 4010 | 9087724 |