Gene Transcripts

Gene Symbol
Pip4k2a
Entrez Gene ID
18718
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
Gene Aliases
AW742916, Pip5k2a
Chromosome Location
Chr. 2: 18842255 - 18998527 on Build GRCm38
UniGene ID
Mm.313977
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008845.4 NP_032871.3 4 529
GenBank mRNA AB009615.1 - 4 454
AK012196.1 - 4 532
AK040558.1 - 4 419
BC011097.1 - 4 476 3672732

View Full Product Details