Gene Transcripts
- Gene Symbol
- Pkdrej
- Entrez Gene ID
- 18766
- Gene Name
- polycystic kidney disease (polycystin) and REJ (sperm receptor for egg jelly homolog, sea urchin)
- Gene Aliases
- Chromosome Location
- Chr. 15: 85814676 - 85821733 on Build GRCm38
- UniGene ID
- Mm.89965
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011105.2 |
NP_035235.2
|
1 |
1349 |
|
View Full Product Details