Gene Transcripts
- Gene Symbol
- Smarcc1
- Entrez Gene ID
- 20588
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1
- Gene Aliases
- AI115498, BAF155, Rsc8, SRG3
- Chromosome Location
- Chr. 9: 110132024 - 110240178 on Build GRCm38
- UniGene ID
- Mm.85410
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009211.2 |
NP_033237.2
|
5, 6 |
677 |
|
GenBank mRNA |
AK042194.1 |
- |
6, 7 |
590 |
|
AK133922.1 |
- |
5, 6 |
652 |
|
AK143485.1 |
- |
5, 6 |
665 |
|
AK144122.1 |
- |
5, 6 |
651 |
|
AK144631.1 |
- |
5, 6 |
645 |
|
AK145352.1 |
- |
5, 6 |
644 |
|
BC022173.1 |
- |
5, 6 |
641 |
|
BC052423.1 |
- |
5, 6 |
639 |
6400646 |
BC053064.1 |
- |
5, 6 |
633 |
|
BC056375.1 |
- |
5, 6 |
640 |
|
U85614.1 |
- |
5, 6 |
638 |
|
View Full Product Details