Gene Transcripts
- Gene Symbol
- Pik3cg
- Entrez Gene ID
- 30955
- Gene Name
- phosphoinositide-3-kinase, catalytic, gamma polypeptide
- Gene Aliases
- 5830428L06Rik, PI3Kgamma, p110gamma
- Chromosome Location
- Chr. 12: 32173129 - 32208677 on Build GRCm38
- UniGene ID
- Mm.101369
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146200.1 |
NP_001139672.1
|
2, 3 |
2137 |
|
NM_001146201.1 |
NP_001139673.1
|
2, 3 |
2193 |
|
NM_020272.2 |
NP_064668.2
|
2, 3 |
2314 |
|
GenBank mRNA |
AF208345.1 |
- |
1, 2 |
1993 |
|
AJ249280.1 |
- |
2, 3 |
2314 |
|
AK040543.1 |
- |
2, 3 |
2099 |
|
AK132553.1 |
- |
2, 3 |
2139 |
|
AK137470.1 |
- |
2, 3 |
2046 |
|
AK137620.1 |
- |
2, 3 |
2195 |
|
BC051246.1 |
- |
2, 3 |
2054 |
6514979 |
View Full Product Details