Gene Transcripts

Gene Symbol
Mysm1
Entrez Gene ID
320713
Gene Name
myb-like, SWIRM and MPN domains 1
Gene Aliases
C130067A03Rik, C530050H10Rik
Chromosome Location
Chr. 4: 94942037 - 94979302 on Build GRCm38
UniGene ID
Mm.208868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177239.2 NP_796213.2 20 2477
GenBank mRNA AK173300.1 - 17 2219
BC150946.1 - 20 2473 9087857
BC151172.1 - 20 2473 9088061

View Full Product Details