Gene Transcripts

Gene Symbol
Nlrp9c
Entrez Gene ID
330490
Gene Name
NLR family, pyrin domain containing 9C
Gene Aliases
AA636537, BB062542, Nalp-zeta, Nalp9c
Chromosome Location
Chr. 7: 26364702 - 26403682 on Build GRCm38
UniGene ID
Mm.212174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042612.1 NP_001036077.1 2 822
GenBank mRNA AK145425.1 - 1 435
AK163321.1 - 2 590
AY360472.1 - 817
AY596203.1 - 2 822

View Full Product Details