Gene Transcripts

Gene Symbol
Wdr95
Entrez Gene ID
381693
Gene Name
WD40 repeat domain 95
Gene Aliases
4930434E21Rik
Chromosome Location
Chr. 5: 149528679 - 149611894 on Build GRCm38
UniGene ID
Mm.136919
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029440.3 NP_083716.2 15, 16 1360
GenBank mRNA AK015309.1 - 13, 14 1464
AK087389.1 - 15, 16 1360

View Full Product Details