Gene Transcripts

Gene Symbol
Plrg1
Entrez Gene ID
53317
Gene Name
pleiotropic regulator 1, PRL1 homolog (Arabidopsis)
Gene Aliases
AA958940, C80566, Tango4
Chromosome Location
Chr. 3: 83055538 - 83072291 on Build GRCm38
UniGene ID
Mm.286349
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016784.3 NP_058064.2 13 1339
GenBank mRNA AF044334.1 - 12 1231
AK075993.1 - 13 1327
AK076007.1 - 13 1317
AK077197.1 - 13 1340
AK080068.1 - 13 1320
BC006750.1 - 13 1309 3709134
BC052895.1 - 13 1297

View Full Product Details