Gene Transcripts

Gene Symbol
Nudt16l1
Entrez Gene ID
66911
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1
Gene Aliases
1110001K21Rik, 5330437I08Rik, Sdos
Chromosome Location
Chr. 16: 4939111 - 4941021 on Build GRCm38
UniGene ID
Mm.428698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025839.3 NP_080115.1 2 303
GenBank mRNA AF435792.1 - 2 315
AK002378.1 - 2 296
AK003241.1 - 2 304
AK007897.1 - 2 299
AK019936.1 - 2 306
AK159841.1 - 2 296
AK186853.1 - 1 13
AK188290.1 - 1 13
BC025569.1 - 2 302 5322456
BC028494.1 - 2 296 1511165

Gene Symbol
Gm16861
Entrez Gene ID
100503704
Gene Name
predicted gene, 16861
Gene Aliases
Chromosome Location
Chr. 16: 4935164 - 4939585 on Build GRCm38
UniGene ID
Mm.480572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK078615.2 - 1 87
BC061511.1 - 1 133

View Full Product Details