Gene Transcripts
- Gene Symbol
- Rusc1
- Entrez Gene ID
- 72296
- Gene Name
- RUN and SH3 domain containing 1
- Gene Aliases
- 2210403N08Rik, AA408288, NESCA
- Chromosome Location
- Chr. 3: 89083975 - 89093363 on Build GRCm38
- UniGene ID
- Mm.27687
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083807.1 |
NP_001077276.1
|
7, 8 |
2031 |
|
NM_001083808.1 |
NP_001077277.1
|
6, 7 |
571 |
|
NM_028188.2 |
NP_082464.2
|
8, 9 |
2442 |
|
GenBank mRNA |
AK032334.1 |
- |
6, 7 |
1061 |
|
AK039664.1 |
- |
8, 9 |
2370 |
|
AK042689.1 |
- |
7, 8 |
1979 |
|
AK170012.1 |
- |
6, 7 |
585 |
|
AK170210.1 |
- |
6, 7 |
534 |
|
AK180118.1 |
- |
3, 4 |
205 |
|
BC052081.1 |
- |
5, 6 |
765 |
|
BC056360.1 |
- |
7, 8 |
1969 |
6816267 |
BC057034.1 |
- |
6, 7 |
637 |
5687517 |
View Full Product Details