Gene Transcripts

Gene Symbol
Atp11b
Entrez Gene ID
76295
Gene Name
ATPase, class VI, type 11B
Gene Aliases
1110019I14Rik, ATPIF, ATPIR, mKIAA0956
Chromosome Location
Chr. 3: 35754062 - 35856276 on Build GRCm38
UniGene ID
Mm.239416
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029570.3 NP_083846.2 2 324
GenBank mRNA AK129248.1 - 2 316
AK154156.1 - 2 328
AK155372.1 - 2 328
BC076603.1 - 2 194 30629594
CN535194.1 - 2 324

View Full Product Details