Gene Transcripts

Gene Symbol
Limch1
Entrez Gene ID
77569
Gene Name
LIM and calponin homology domains 1
Gene Aliases
3732412D22Rik, mKIAA1102
Chromosome Location
Chr. 5: 66745840 - 67057159 on Build GRCm38
UniGene ID
Mm.374778
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001980.2 NP_001001980.2 6, 7 557
GenBank mRNA CD804084.1 - 1, 2 17
CF540375.1 - 4, 5 308

View Full Product Details