Gene Transcripts

Gene Symbol
Inpp1
Entrez Gene ID
16329
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
2300002C06Rik, AV137389
Chromosome Location
Chr. 1: 52785425 - 52817688 on Build GRCm38
UniGene ID
Mm.917
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008384.2 NP_032410.2 4 655
GenBank mRNA AK078382.1 - 4 543
BC025072.1 - 4 400 4456812
U27295.1 - 4 504

View Full Product Details