Gene Transcripts

Gene Symbol
Trp53i13
Entrez Gene ID
216964
Gene Name
transformation related protein 53 inducible protein 13
Gene Aliases
2410019G02Rik, Tp53i13
Chromosome Location
Chr. 11: 77508099 - 77515921 on Build GRCm38
UniGene ID
Mm.341211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024920.1 NP_001020091.1 6 748
GenBank mRNA BC147324.1 - 6 746
BC147325.1 - 6 746 9056377

View Full Product Details