Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285826.1 | NP_001272755.1 | 6 | 976 | |
NM_001285827.1 | NP_001272756.1 | 6 | 641 | ||
NM_027351.3 | NP_081627.1 | 6 | 455 | ||
NM_027374.3 | NP_081650.2 | 6 | 641 | ||
GenBank mRNA | AF315803.1 | - | 6 | 589 | |
AK010207.1 | - | 6 | 459 | ||
AK011000.1 | - | 6 | 410 | ||
BC060993.1 | - | 6 | 423 | 6591278 | |
BC061645.1 | - | 6 | 919 | 5032617 | |
DV043111.1 | - | 6 | 677 |