Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029934.3 | NP_084210.2 | 5 | 1158 | |
GenBank mRNA | AB055410.1 | - | 5 | 1156 | |
AK019981.1 | - | 5 | 569 | ||
AK038998.1 | - | 5 | 580 | ||
AK044149.1 | - | 5 | 585 | ||
AK150010.1 | - | 5 | 552 | ||
AK154606.1 | - | 5 | 1157 | ||
AK158998.1 | - | 4 | 913 | ||
AK170124.1 | - | 5 | 635 | ||
BC023417.1 | - | 5 | 606 | 5028759 | |
BC118958.2 | - | 5 | 803 | 40093821 | |
EU016380.1 | - | 4 | 370 |