Gene Transcripts

Gene Symbol
Psg22
Entrez Gene ID
243862
Gene Name
pregnancy-specific glycoprotein 22
Gene Aliases
AA408604, cea9
Chromosome Location
Chr. 7: 18718090 - 18727248 on Build GRCm38
UniGene ID
Mm.389833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004152.2 NP_001004152.1 3 916
GenBank mRNA BC050099.1 - 3 816
BC080694.1 - 3 928 30433272

View Full Product Details