Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134095.2 | NP_598856.1 | 5 | 500 | |
GenBank mRNA | AK005706.1 | - | 5 | 491 | |
AK005963.1 | - | 5 | 456 | ||
AK016347.1 | - | 5 | 408 | ||
AK131703.1 | - | 5 | 454 | ||
AK146155.1 | - | 5 | 524 | ||
AK170731.1 | - | 6 | 769 | ||
AK189878.1 | - | 5 | 258 | ||
BC022097.1 | - | 4 | 455 | ||
BC056972.1 | - | 5 | 500 | 6811343 | |
BC071205.1 | - | 5 | 392 | 6515068 | |
BC131979.1 | - | 5 | 396 | 40130256 | |
BC132642.1 | - | 5 | 396 | 40130919 | |
BC145235.1 | - | 4 | 326 |