Gene Transcripts
- Gene Symbol
- Otub2
- Entrez Gene ID
- 68149
- Gene Name
- OTU domain, ubiquitin aldehyde binding 2
- Gene Aliases
- 2010015L18Rik, 4930586I02Rik, AI413508, AW557219, OTB2, OTU2
- Chromosome Location
- Chr. 12: 103376894 - 103406351 on Build GRCm38
- UniGene ID
- Mm.127378
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177841.1 |
NP_001171312.1
|
4, 5 |
839 |
|
NM_026580.4 |
NP_080856.1
|
3, 4 |
413 |
|
GenBank mRNA |
AK006054.1 |
- |
3, 4 |
335 |
|
AK006346.1 |
- |
3, 4 |
365 |
|
AK006469.1 |
- |
3, 4 |
362 |
|
AK008258.2 |
- |
3, 4 |
297 |
|
AK016449.1 |
- |
3, 4 |
397 |
|
AK019830.1 |
- |
3, 4 |
390 |
|
AK076336.1 |
- |
4, 5 |
842 |
|
AK158758.1 |
- |
3, 4 |
696 |
|
AK160505.1 |
- |
3, 4 |
392 |
|
BC049551.1 |
- |
3, 4 |
396 |
6771956 |
BC052409.1 |
- |
3, 4 |
328 |
|
View Full Product Details