Gene Transcripts

Gene Symbol
Stk38
Entrez Gene ID
106504
Gene Name
serine/threonine kinase 38
Gene Aliases
5830476G13Rik, 9530097A09Rik, AA617404, Ndr1
Chromosome Location
Chr. 17: 28970885 - 29008006 on Build GRCm38
UniGene ID
Mm.439986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134115.2 NP_598876.1 13 1415
GenBank mRNA AY292399.1 - 13 1279
BC009658.1 - 13 1415 3484764

View Full Product Details