Gene Transcripts

Gene Symbol
Laptm4b
Entrez Gene ID
114128
Gene Name
lysosomal-associated protein transmembrane 4B
Gene Aliases
C330023P13Rik, LAPTM4beta
Chromosome Location
Chr. 15: 34238026 - 34284295 on Build GRCm38
UniGene ID
Mm.197518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033521.3 NP_277056.1 5 754
GenBank mRNA AF317418.1 - 5 456
AK077269.1 - 5 617
AK077933.1 - 5 754
AK079581.1 - 5 617
AK082807.1 - 5 633
AK146868.1 - 4 520
AK166014.1 - 5 656
BC019120.1 - 5 624 4507607
EU561010.1 - 5 754

View Full Product Details