Gene Transcripts

Gene Symbol
Laptm4a
Entrez Gene ID
17775
Gene Name
lysosomal-associated protein transmembrane 4A
Gene Aliases
AA286466, LAPTM4, MTP, Mtrp, mKIAA0108
Chromosome Location
Chr. 12: 8921307 - 8938742 on Build GRCm38
UniGene ID
Mm.30071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008640.2 NP_032666.2 5 1351
GenBank mRNA AK013963.1 - 4 369
AK075699.1 - 5 586
AK084515.1 - 5 994
AK088958.1 - 5 611
AK165638.1 - 5 620
BC086667.1 - 5 531 30633914
BC089021.1 - 5 564 6734038
U34259.1 - 6 1218

View Full Product Details