Gene Transcripts

Gene Symbol
Myog
Entrez Gene ID
17928
Gene Name
myogenin
Gene Aliases
MYF4, bHLHc3, myo
Chromosome Location
Chr. 1: 134290004 - 134292548 on Build GRCm38
UniGene ID
Mm.16528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031189.2 NP_112466.1 1 328
GenBank mRNA AB221643.1 - 1 276
BC048683.1 - 1 343 6516364
BC068019.1 - 1 309 6508229
CJ096370.1 - 1 328
D90156.1 - 1 319
X15784.1 - 1 324

View Full Product Details