Gene Transcripts

Gene Symbol
Rfxank
Entrez Gene ID
19727
Gene Name
regulatory factor X-associated ankyrin-containing protein
Gene Aliases
Tvl1
Chromosome Location
Chr. 8: 70130806 - 70139197 on Build GRCm38
UniGene ID
Mm.161167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025589.1 NP_001020760.1 2 328
NM_011266.2 NP_035396.1 2 328
GenBank mRNA AF094761.1 - 2 244
AF123704.1 - 2 229
AK052298.1 - 2 285
AK087462.1 - 2 257
AK146148.1 - 2 257
BC010971.1 - 2 262 4190691
CN526109.1 - 2 328
L43164.1 - 2 229

View Full Product Details