Gene Transcripts

Gene Symbol
Sstr3
Entrez Gene ID
20607
Gene Name
somatostatin receptor 3
Gene Aliases
Smstr-3, Smstr3, sst3
Chromosome Location
Chr. 15: 78537014 - 78549369 on Build GRCm38
UniGene ID
Mm.211411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009218.3 NP_033244.2 3 796
GenBank mRNA AK136904.1 - 3 797
BC120843.1 - 1 439 40129766
BC137670.1 - 1 439 8860689

View Full Product Details