Gene Transcripts

Gene Symbol
Nckap5
Entrez Gene ID
210356
Gene Name
NCK-associated protein 5
Gene Aliases
8430408F21, D130011D22Rik, E030049G20Rik, Erih1, Erih2, Gm1548, Nap5
Chromosome Location
Chr. 1: 125913636 - 126830897 on Build GRCm38
UniGene ID
Mm.377234
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081756.1 NP_001075225.1 10 1111
NM_172484.3 NP_766072.2 8 972
NM_176957.3 NP_795931.2 10 1111
GenBank mRNA AK053243.1 - 8 794
AY957601.1 - 6 531
AY957602.1 - 6 531
BC066095.1 - 8 972 6856184
BC079551.1 - 8 972 6856184

View Full Product Details