Gene Transcripts

Gene Symbol
Tgfbr2
Entrez Gene ID
21813
Gene Name
transforming growth factor, beta receptor II
Gene Aliases
1110020H15Rik, AU042018, DNIIR, RIIDN, TBR-II, TbetaR-II, TbetaRII
Chromosome Location
Chr. 9: 116087695 - 116175363 on Build GRCm38
UniGene ID
Mm.172346
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009371.3 NP_033397.3 6 1690
NM_029575.3 NP_083851.3 5 1615
GenBank mRNA AF406755.1 - 5 1289
AK043619.1 - 6 1774
AK046561.1 - 6 1616
AK053793.1 - 6 1691
AK148167.1 - 5 1615
AK159327.1 - 6 1670
BC052629.1 - 1592
S69114.1 - 5 1326

View Full Product Details