Gene Transcripts

Gene Symbol
Rfesd
Entrez Gene ID
218341
Gene Name
Rieske (Fe-S) domain containing
Gene Aliases
AI256775, D030068K09
Chromosome Location
Chr. 13: 76001535 - 76018712 on Build GRCm38
UniGene ID
Mm.266515
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131068.1 NP_001124540.1 4 981
NM_001131069.1 NP_001124541.1 4 444
NM_178916.5 NP_849247.3 4 1431
GenBank mRNA AK040255.1 - 4 936
AK083705.1 - 4 1432
BC030381.1 - 4 1056 5375753
CN839985.1 - 4 454

View Full Product Details