Gene Transcripts

Gene Symbol
Gpbar1
Entrez Gene ID
227289
Gene Name
G protein-coupled bile acid receptor 1
Gene Aliases
BG37, GPCR, GPR131, M-BAR, TGR5
Chromosome Location
Chr. 1: 74278600 - 74279589 on Build GRCm38
UniGene ID
Mm.246587
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174985.1 NP_778150.1 1 330
GenBank mRNA AB086171.1 - 1 330
AB089308.1 - 1 330
BC116914.1 - 1 380 40126233
BC119327.1 - 1 380 8734076

View Full Product Details