Gene Transcripts

Gene Symbol
Klk15
Entrez Gene ID
317652
Gene Name
kallikrein related-peptidase 15
Gene Aliases
Chromosome Location
Chr. 7: 43929405 - 43939852 on Build GRCm38
UniGene ID
Mm.389896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174865.1 NP_777354.1 2 134
GenBank mRNA BC106965.1 - 2 134
BC106966.2 - 2 131
BC107012.1 - 2 134 40047687

View Full Product Details