Gene Transcripts

Gene Symbol
Pkd2l1
Entrez Gene ID
329064
Gene Name
polycystic kidney disease 2-like 1
Gene Aliases
B830002B15, BC046386, PCL, PKD2L, Pkdl, TRPP3
Chromosome Location
Chr. 19: 44147637 - 44248354 on Build GRCm38
UniGene ID
Mm.308481
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181422.3 NP_852087.2 15 2426
GenBank mRNA AB290927.1 - 15 2258
AF271381.1 - 15 2392
AK046772.1 - 14 2636

View Full Product Details