Gene Transcripts

Gene Symbol
Rhox1
Entrez Gene ID
385343
Gene Name
reproductive homeobox 1
Gene Aliases
Chromosome Location
Chr. X: 37213804 - 37222258 on Build GRCm38
UniGene ID
Mm.379566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025084.2 NP_001020255.2 1 59
GenBank mRNA DQ058638.1 - 1 59

View Full Product Details