Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015822.2 | NP_056637.1 | 2 | 674 | |
GenBank mRNA | AF176521.1 | - | 2 | 381 | |
AK080863.1 | - | 2 | 683 | ||
AK142007.1 | - | 2 | 674 | ||
AK166128.1 | - | 2 | 555 | ||
AK167409.1 | - | 2 | 493 | ||
BC026835.1 | - | 1 | 340 | ||
BC037665.1 | - | 1 | 337 | ||
BC067203.1 | - | 2 | 556 | 6334317 |